Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Flush Seal Drain Valve, Ace Glass Incorporated
Supplier: Ace Glass
6472-245
89054-660EA 2194.6 USD InStock
89054-660
Flush Seal Drain Valve, Ace Glass Incorporated
Valves Line Valves
Flush seal drain assembly with PTFE valve stem and PFA encapsulated silicone O-Ring for use with beaded reaction flasks
By rotating the handle, plug rises into the flask and seals to afford zero dead space. Back thread out to drain through 1-inch beaded pipe angled side port. Supplied with CAPFE O-Ring for seal at flask, two Kalrez O-Rings supplied on valve stem. For stabilization, when using long shaft lengths or stirring viscous media, it is suggested that the flush seal drain valve with pin be used. This will prevent shaft oscillation or vibration and help to maintain a concentric stirring motion.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty