Would you like to log in again?
Pilot Plant Reactor Support Stand, Ace Glass Incorporated
Supplier: Ace Glass
6473-47 6472-360 6472-328 6472-335 6473-36 6473-34 6472-331 6472-332 6473-51 6473-37 6473-35 6472-337 6472-329
89054-690EA 6005.21 USD InStock
89054-690 89084-670 89084-668 89054-832 89054-700 89054-834 89054-836 89054-718 89054-838 89054-694 89054-696 89054-688 89054-698
Pilot Plant Reactor Support Stand, Ace Glass Incorporated
Support Stands
Four post stainless steel pilot plant reactor stand with epoxy coated pipe connections.
Removable side shelf add-on for ACE pilot plant reactor support stands. Suitable for placement of motor controllers, temperature controllers, relay units. The black powder-coated aluminum plate has a 3/8" wide x 3/8" high ledge to prevent items from falling or sliding off. Attaches to stand with galvanized iron clamp-on tee with set screws and backing bolt.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty