Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Beaded Pipe Drain Valve Assembly, Ace Glass Incorporated
Supplier: Ace Glass
6472-99
89054-802EA 908.45 USD InStock
89054-802
Beaded Pipe Drain Valve Assembly, Ace Glass Incorporated
Valves Line Valves
This flask is supplied with a 1" beaded pipe-type flange extension off bottom for connection to a 0–20 mm threaded PTFE stopcock as the outlet
Stopcock assembly has a mating 1" beaded flange that connects to extension off bottom via a Viton lined coupling. Takeoff on stopcock is 1" O.D. tubing. Drain assembly plug supplied with FETFE o-rings; Chemraz® o-rings can be offered as replacements. ACE can supply the takeoff with beaded flange, Ace-Thred, hose connection. Listed as flask with beaded pipe extension only, drain assembly only (includes 0-20 plug plus glass thread with mating beaded pipe) or flask with drain assembly. Order coupling separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty