Would you like to log in again?
Reaction Flask, Conical 4-Inch Flange, Ace Glass Incorporated
Supplier: Ace Glass
6476-05 6476-25 6476-15 6476-10 6476-20
89054-900EA 276.25 USD InStock
89054-900 89054-946 89054-992 89054-884 89054-886
Reaction Flask, Conical 4-Inch Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Rugged cylindrical flask with conical neck opening of four inches (100mm)
Uses a clamp for securing flask head to the flask. Fabricated from borosilicate glass.
Ordering information: For top PTFE gasket, order 89055-004.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty