Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Reaction Flask with Detached Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
6492-02 6492-25 6492-06 6492-10 6492-15
89054-986EA 377.53 USD InStock
89054-986 89054-988 89054-990 89054-906 89054-982
Reaction Flask with Detached Stopcock, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This reaction flask has a detached 4mm bore glass stopcock
The flask is separated from the stopcock and outlet tube by a [SJ] 28/15 joint, ball on the flask, socket on the stopcock. Fabricated from borosilicate glass.
Ordering information: Complete apparatus includes flask and stopcock. For bottom joint clamp (not included), order 89060-256. For a clamp (not included) to secure the 4" head of your choice (not included) to the flask, order 89055-014.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty