Would you like to log in again?
Clamp, Two-Piece, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6508-11 6508-06
89055-038EA 491.17 USD InStock
89055-038 89055-040
Clamp, Two-Piece, Ace Glass Incorporated
Clamps General Purpose Clamps
Positive, two-piece anodized clamp for securing reaction flasks with flat-style flanges
Black anodized, high strength aluminum clamp has three wing nuts to secure the bottom member to the flask head. Simply loosen the wing nut and the nut tilts free from the upper member.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty