Would you like to log in again?
Flask, Reaction, Flat Flange, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6511-08 6511-12 6511-10 6511-06
89055-056EA 507.63 USD InStock
89055-056 89055-054 89055-052 89055-050
Flask, Reaction, Flat Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Cylindrical, heavy-wall reaction flask, round bottom, with flat, ground flange
Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Fabricated from borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty