Would you like to log in again?
Flask, Reaction, Flat Flange, Flat Bottom, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6511-27 6511-31 6511-29 6511-24
89055-058EA 344.23 USD InStock
89055-058 89055-064 89055-062 89055-060
Flask, Reaction, Flat Flange, Flat Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Cylindrical heavy wall reaction flask, with flat bottom rounded into side wall, and flat, ground flange top
Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Fabricated from borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty