Would you like to log in again?
Quick Release Clamps, Stainless Steel, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6517-24 6517-54 6517-60 6517-31 6517-27 6517-56 6517-25 6517-22
89055-114EA 343.49 USD InStock
89055-114 89055-112 89055-110 89055-120 89055-108 89055-118 89055-106 89055-116
Quick Release Clamps, Stainless Steel, Ace Glass Incorporated
Clamps General Purpose Clamps
Sturdy, stainless steel, quick release clamps can be used with two-piece pressure reaction flasks/heads that feature Duran flanges
Multiple sizes available with or without an extension rod (for clamping to a support frame).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty