Would you like to log in again?
Unjacketed Cylindrical Reaction Flasks with Valves, Duran Flange, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6522-85 6522-13 6522-11 6522-86 6522-17 6522-84 6522-15 6522-81 6522-82 6522-83 6472-02
89055-202EA 11316.15 USD InStock
89055-202 89054-568 89055-200 89055-144 89055-198 89055-142 89055-196 89055-140 89055-194 89055-138 89055-204
Unjacketed Cylindrical Reaction Flasks with Valves, Duran Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These unjacketed cylindrical reaction flask are made of heavy wall borosilicate glass and include a bottom outlet valve. Each flask has high visibility capacity reference graduations for easy viewing. They can be used standalone or with bottom opening heating mantles. Available with Duran O-ring groove or KF Plane flat ground flanges. as well as a variety of drain valve styles to select from.
Reactor accessories including Inlet and Outlet Connections, Duran Flange O-Rings PFA Encapsulated Silicone, Duran Quick Release Clamps, Spherical Joint Adapters for Valves, Couplings, Spherical Joint O-Rings, Bottom Outlet Valves, and Beaded Pipe Couplings for Valves are sold separately.
Each flask has high visibility capacity reference graduations for easy viewing. They can be used standalone or with bottom opening heating mantles. Available with Duran O-ring groove or KF Plane flat ground flanges. as well as a variety of drain valve styles to select from.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty