Would you like to log in again?
Jacketed Cylindrical Reaction Flask with Duran Flange, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6524-320 6524-317 6523-316 6524-20 6523-12 6523-314 6523-27 6523-319 6524-313 6523-312 6523-30 6524-17 6524-315 6523-37 6523-19 6523-22 6523-32 6524-15 6523-14 6524-13 6523-16
89055-214EA 5251.26 USD InStock
89055-214 89055-224 89055-212 89055-222 89055-210 89055-220 89055-208 89055-218 89055-228 89055-206 89055-216 89055-226 10767-296 10767-294 10767-178 89083-550 10767-176 10767-298 10767-292 10767-180 10767-290
Jacketed Cylindrical Reaction Flask with Duran Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
The rugged cylindrical reaction flask with Duran style flat o-ring groove flange, is for use with quick release clamp. Fabricated from heavy wall borosilicate glass. Inlet and outlet connections on jacket, sealed tangentially, are o-ring ball joints. Small reactors 1 to 6 L use 28/15 spherical joints and 10 L and larger us 35/25 o-ring joints. Ten liter and larger are supplied with capacity reference graduations. Each flask is supplied with one CAPFE, PTFE encapsulated silicone, o-ring. Flasks are available with a variety of valve types. Maximum Jacket Pressure: 5 PSI (.34 Bar). Jacket Connections: 35/25 (S/J). Maximum Delta T: 60°C.
Fabricated from heavy wall borosilicate glass. Inlet and outlet connections on jacket, sealed tangentially, are o-ring ball joints. Small reactors 1 to 6 L use 28/15 spherical joints and 10 L and larger us 35/25 o-ring joints. Ten liter and larger are supplied with capacity reference graduations. Each flask is supplied with one CAPFE, PTFE encapsulated silicone, o-ring. Flasks are available with a variety of valve types. Maximum Jacket Pressure: 5 PSI (.34 Bar). Jacket Connections: 35/25 (S/J). Maximum Delta T: 60°C. Easy Action Bottom Valve: Flask has Easy Action Plug bottom outlet with Chemraz® o-rings for draining contents. Bottom outlet has size 35/25 o-ring ball joint connection. Supplied with one CAPFE o-ring. Sink Type Valve: Flask is fabricated with a #36 Ace-Thred on bottom. The all PTFE valve is inserted into the Ace-Thred from inside the flask. A stainless steel handle is then secured to the drain tube. By rotating the handle, plug rises into the flask and contents drain through three (3) 1/2" wide x 1 1/2" long slit openings. Rotate handle in opposite direction and plug seals at bottom of flask, with NO dead space. Supplied with two Chemraz® o-rings: one to secure valve in #36 Ace-Thred; the other, on bottom of plug to make seal at bottom of flask.
Piston Type Valve: Flask has 35/25 o-ring ball joint on outlet and utilizes a sealed-in threaded stopcock assembly that rotates closed into the bottom of the flask thereby offering nearly zero dead space. Turn stopcock handle counterclockwise and piston plunger retracts to allow contents to drain. Drain opening is approximately 15 mm. Piston plunger supplied with FETFE o-rings.
Beaded: Flask is supplied with a one inch beaded pipe-type flange extension off bottom for connection to a 0–20 mm threaded stopcock as the outlet. Stopcock assembly has a mating one inch beaded flange that connects to extension off bottom via a Viton lined coupling. Takeoff on stopcock is one inch O.D. tubing. Drain assembly plug supplied with FETFE o-ring.
Flush Seal Valve: Flask is fabricated with a two inch beaded valve seat, for 30 and 50 L. The plug, with a CAPFE o-ring, is inserted into valve seat and secures within a coupling. By rotating the handle, plug rises into the flask and seals to afford zero dead space. Back thread out to drain through one inch beaded pipe angled side port. Supplied with CAPFE o-ring for seal at flask, two Kalrez o-rings supplied on valve stem.
Valves and accessories sold separately. Easy Action and Piston Type Flasks Supplied with Valve.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty