Would you like to log in again?
Vigreux Distilling Column, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6578-30 6578-12 9345-09 9345-10 6578-04 6578-16 6578-24 6578-08 6578-34 6578-10 9345-13 6578-20 6578-22 6578-06 9345-08 9345-11 6578-14 6578-32
89055-664EA 301.2 USD InStock
89055-664 76214-398 89074-422 89074-420 89055-662 76214-402 89055-660 89074-424 76214-400 89074-418 89074-416 89055-658 89055-668 89055-656 89055-666 76214-406 76214-404 76214-408
Vigreux Distilling Column, Ace Glass
Distillation Components Distillation Columns
The column length in millimeters (listed below) refers to the effective length of the column as measured from the lowest to the highest indent of the column
Outer [ST] joint at top, ground [ST] inner joint at bottom. Full-sized joints (only) are tooled with reinforcement rings for added strength and stability. Fabricated from borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty