Would you like to log in again?
Drying Apparatus, Modified Abderhalden, Ace Glass
Supplier: Ace Glass
6692-02 6692-10 6692-04
89055-752EA 1002 USD InStock
89055-752 89055-750 89055-760
Drying Apparatus, Modified Abderhalden, Ace Glass
Desiccators Desiccators, Glass
This modified Abderhalden vacuum drying apparatus features a desiccant tube with two-way action that permits the use of a smaller desiccant volume and better air contact with desiccant
During the final stage of drying, the T-bore stopcock is turned so that the evacuation path is directly over the desiccant, while in the early stages the desiccant is bypassed. The condenser is placed above the boiling flask, but off to the side of the heating tube. The upper [ST] 24/40 joint is offset so that the condensed liquid does not drop over the inner tube. The opening to the inside chamber is [ST] 40/35. Fabricated from borosilicate glass.
Ordering information: Complete item includes 89055-760 drying chamber, [ST] 40/35 joint; 89055-750 desiccant tube, [ST] 40/35; 89053-914 West Condenser (300mm length), [ST] 24/40; and 89056-236 flask (250mL), [ST] 24/40. Individual items may be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty