Would you like to log in again?
Tubes, Freeze Drying with Screw Cap, Ace Glass
Supplier: Ace Glass
6696-31 6696-32
89055-784EA 40.49 USD InStock
89055-784 89055-782
Tubes, Freeze Drying with Screw Cap, Ace Glass
Tubes Culture Tubes
These modified culture tubes come complete with a PTFE lined screw-cap and stops on side to prevent 'suck in' under vacuum.
Both sizes fit inside a #25 Ace-thred bushing for use with 89055-766 'umbrella' manifold. Fabricated from borosilicate glass.
Ordering information: Working capacity of tubes is about one half of the capacity listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty