Would you like to log in again?
Air Sampling Bleed Adapter, Ace Glass Incorporated
Supplier: Ace Glass
7499-23 7499-21 7499-15 7499-12 7499-33
89059-200EA 171.32 USD InStock
89059-200 89059-198 89059-196 89052-502 89055-802
Air Sampling Bleed Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The adjustable bleed adapter is designed to reduce air flow in the air sampling manifold to prevent a pressure drop across the sampling ports.
  • Install in front of blower
  • Controls volume of air flow
  • Fabricated from borosilicate glass
The device is normally installed in front of the blower, in line with the manifold. The positioning of the holes in the threaded nylon plug controls the volume of air flow.
Ordering information: Available complete with bushing (89059-200) and size -220 FETFE O-Ring (89061-028) for 25mm top port. Individual components may also be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty