Would you like to log in again?
Trap, Rotary Evaporator, Anti-Splash, Ace Glass Incorporated
Supplier: Ace Glass
6705-10 6705-26 6705-22 6705-14 6705-12 6705-06
89055-874EA 233.99 USD InStock
89055-874 89055-882 89055-880 89085-004 89055-878 89055-876
Trap, Rotary Evaporator, Anti-Splash, Ace Glass Incorporated
Vacuum Traps
This rotary evaporator trap is designed to greatly reduce carry-over from foaming or bumping.
Two drain holes, close to base of the inner tube, allow solvent to drain back into evaporator flask. Fabricated from borosilicate glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty