Would you like to log in again?
Kriel Photochemical Reactor, Ace Glass Incorporated
Supplier: Ace Glass
6962-35 6962-32 6962-65 6962-62
89057-284EA 3527.55 USD InStock
89057-284 89057-282 89057-286 89057-280
Kriel Photochemical Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
This is a tapered wall style reaction vessel for prep-scale photolysis
Tapered sides assure that the effective area of a 450w or 550w photochemical lamp can be entirely submersed into the liquid, contrary to traditional round bottom reaction flasks. Net result is approximately a 20% savings in reaction time. An additional advantage of this design is in the initial mixing of reactants. The flat bottom allows immediate stirring with a magnetic stirrer and after only one-third full, mechanical stirring can be implemented. Center neck is #50 Ace-Thred for use with 450 mm Immersion Wells. Threaded design offers convenience of vertical depth positioning of well to suit your needs. Two side joints are standard taper 29/42, one for 10 mm stirring shaft and bearing, the other for charging flask or condenser. Two front ports are #7 Ace-Threds, one with 7 mm I.D. bushing for a thermometer, the other with 8 mm I.D. bushing for sparger tube.
Ordering information: Complete unit consists of flask, immersion well (quartz), glass stopper, stirring shaft, bearing, one #7 nylon bushing with 7 mm I.D. hole, one #7 bushing with 8 mm I.D. hole and PTFE stir bar.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty