Would you like to log in again?
Bottle, Filtering, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6989-15 6989-24 6989-21 6989-27 6989-127 6989-118 6989-121 6989-124 6989-115 6989-18
89057-408EA 1199.6 USD InStock
89057-408 89057-410 89057-414 89057-402 89057-412 89057-400 89057-418 89057-406 89057-416 89057-404
Bottle, Filtering, Ace Glass Incorporated
Filters Bottle Top Filters
This heavy wall, bottle shaped, filtering flask with removable polypropylene hose connection is available clear glass or plastic coated.
The plastic coated version are steam autoclavable at 135 °C. Fabricated from borosilicate glass.
Ordering information: For replacement polypropylene hose connection set, order 89057-420.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty