Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Flask, Freeze Drying, Lyophilization, Ace Glass
Supplier: Ace Glass
7035-09 7035-128 7035-46 7035-42 7035-110 7035-52 7035-152 7035-104 7035-17 7035-54 7035-116 7035-122 7035-36 7035-48 7035-56 7035-29 7035-34 7035-21 7035-58 7035-40 7035-27 7035-150 7035-15 7035-160 7035-170 7035-162 7035-03 7035-23 7035-11 7035-05
89057-542EA 458.32 USD InStock
89057-542 89057-520 89057-486 89057-540 89057-484 89057-524 89057-502 89057-500 89057-522 89057-488 89057-506 89057-528 89057-526 89057-504 89057-518 89057-494 89057-492 89057-498 89057-530 89057-496 89057-512 89057-534 89057-532 89057-510 89057-538 89057-516 89057-536 89057-514 89057-508 89057-490
Flask, Freeze Drying, Lyophilization, Ace Glass
Flasks Flat Bottom Flasks
Complete "fast-freeze" flask with snap-on rubber cap and 1/2" or 3/4" O.D.
  • Fabricated from borosilicate glass
  • Flasks are designed for ease of handling and faster loading
Snap-on black rubber cap is grease free; one size fits all capacity flasks. Plain straight adapter for connecting flask cap to lyophilizer manifold is removable and fabricated from PTFE. PTFE adapter with shut-off valve also available. The size value accompanied with adapter size and style.
Plain or valve adapter, compatible with all major brand freeze drying manifold valves. Uniform wall weight maximizes heat transfer while maintaining structural strength. Wide (53 mm I.Ø) mouth provides easy access to inner walls.
Ordering information: All items also available separately.
Delivery information: Available complete with flask, cap and plain straight PTFE adapter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty