Would you like to log in again?
Flask, Volumetric, Class A, Ace Glass Incorporated
Supplier: Ace Glass
7124-20 7124-15
89057-576EA 208.39 USD InStock
89057-576 89053-114
Flask, Volumetric, Class A, Ace Glass Incorporated
Flasks Volumetric Flasks
A specially designed, standardized volumetric flask, with stopper
All capacities have a standard taper 12/18 outer joint with an opening of approximately 10mm. The cylindrical body affords better mixing and drainage, and ease of withdrawal using pipets. The wide base offers greater stability. The upper surface of the base is ground to allow pencil markings. Class “A” tolerances. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty