Would you like to log in again?
Straight Gas Dispersion Tube, Ace Glass Incorporated
Supplier: Ace Glass
7197-14 7197-06 7197-18 7197-12 7197-02 7197-08 7197-16 7197-04
89058-408EA 148.57 USD InStock
89058-408 89058-350 89058-330 89058-352 89058-332 89058-334 89058-314 89058-348
Straight Gas Dispersion Tube, Ace Glass Incorporated
Gas Tubes
The top tube features an 8mm O.D., and the total length of the tube is 150mm
Thickness of the bottom is 14mm. The bottom O.D. matches the disc diameter, and the overall length is 164mm. Available in two different disc diameters, and four different porosities. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty