Would you like to log in again?
Gas Dispersion Tube, 15° Angle, Ace Glass Incorporated
Supplier: Ace Glass
7204-08 7204-07 7204-03 7204-04
89058-380EA 67.42 USD InStock
89058-380 89058-374 89058-376 89058-378
Gas Dispersion Tube, 15° Angle, Ace Glass Incorporated
Gas Tubes
Gas dispersion tube with porous fritted tip for use with #7 Ace-Threds
Tube is 6.5mm O.D. and bent at 15° to allow positioning inside a flask to avoid stirring paddles and yet reach close to bottom of flask. Frit length is 10mm. Fabricated from borosilicate glass.
Ordering information: Available in two different porosities and lengths.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty