Would you like to log in again?
Addition Funnels, Ungraduated with 1:5 PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
7257-50 7257-52 7257-54
89058-644EA 315.88 USD InStock
89058-644 89058-646 89058-648
Addition Funnels, Ungraduated with 1:5 PTFE Plug, Ace Glass Incorporated
Funnels Dropping Funnels
Ungraduated, with a standard taper joint at top and bottom
For stoppers, order 89062-824. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty