Would you like to log in again?
Funnels, Pressure Equalizing, Graduated with Glass Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7296-56 7296-58 7296-52 7296-54 7296-50
89058-726EA 442.39 USD InStock
89058-726 89058-728 89058-730 89058-732 89058-734
Funnels, Pressure Equalizing, Graduated with Glass Stopcock, Ace Glass Incorporated
Funnels Dropping Funnels
Cylindrical with standard taper joint top and bottom, drip stem, pressure equalizing arm and double scale graduations, calibrated from stopcock
Drip stem does not extend beyond joint, thereby eliminating possible source of breakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty