Would you like to log in again?
Addition Funnels, Graduated, Jacketed with PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7270-41 7270-37
89058-772EA 1043.28 USD InStock
89058-772 89058-662
Addition Funnels, Graduated, Jacketed with PTFE Stopcock, Ace Glass Incorporated
Funnels Dropping Funnels
Double scale graduated funnel with jacket for cooling with dry ice
Calibrated up from stopcock. With 1:5 PTFE stopcock plug. Fabricated from borosilicate glass.
Ordering information: For stoppers (not supplied), order 89062-824.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty