Would you like to log in again?
Pressure Equalizing Funnels, Graduated with PTFE Metering Valve, Ace Glass Incorporated
Supplier: Ace Glass
7320-17 7320-10 7320-14 7320-12 7320-08
89058-780EA 617.88 USD InStock
89058-780 89058-782 89058-784 89058-776 89058-778
Pressure Equalizing Funnels, Graduated with PTFE Metering Valve, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical with standard taper joint top and bottom, drip stem, pressure equalizing arm and double scale graduations, calibrated from stopcock
Drip stem does not extend beyond joint, thereby eliminating possible source of breakage. Stopcock available with glass plug, 1:5 PTFE plug or PTFE metering plug. Fabricated from borosilicate glass.
Ordering information: Stoppers are not supplied. For stoppers or replacement stopcock plugs, contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty