Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Gas Manifold, Ace Glass Incorporated
Supplier: Ace Glass
7416-14 7416-10
89058-826EA 870.38 USD InStock
89058-826 89058-824
Gas Manifold, Ace Glass Incorporated
Manifolds
Designed for use as a portable gas analysis apparatus
Manifolds are available with either three or four straight bore 2 mm glass stopcocks and one three-way stopcock with back port, fabricated on capillary tubing. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty