Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogenation/Gas Apparatus I, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7482-38 7482-42 7482-77 7482-31 7482-50 7482-46 7482-37 7482-70 7482-25 7482-33 7482-20 7482-500
89058-902EA 13226.05 USD InStock
89058-902 10767-186 89058-906 89058-890 89058-892 89058-884 89058-894 89058-886 89058-896 89058-888 89058-898 89058-900
Hydrogenation/Gas Apparatus I, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Parr Model 3911 Hydrogenation unit for working volumes of 1 to 200 mL

Modified Vessel Clamp Allows Clamping of 89051-324 Temperature Measurement Adapter to Parr Model 3911 Hydrogenation Units
Nylon Adapter Connects Vessel to Gas Tank Tubing On Smaller #7 Ace-Thred Vessels
PTFE Adapter Connects Vessel to Gas Tank Tubing On The Larger #25 Ace-Thred Vessels
Upper and Lower Bumper Pad Protects Glassware In 89058-884 and 89058-908 Hydrogenation Units

Ace-thred vessels replace rubber stoppered vessels and eliminate cross-over contamination caused by leeching or sample retension. Complete unit includes all adapters needed for bottles (1 - 4 mL, 3 - 8 mL, 8 - 20 mL, 10 - 35 mL, 25 - 75 mL and 50 - 200 mL). Units without vessels and adapters are available in explosion proof and air driven models.

Ordering information: No glassware or adapters included, except for the complete unit (7482-50).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty