Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Air Sampling Filter Screens, Ace Glass Incorporated
Supplier: Ace Glass
7503-05 7503-15 7503-25
89059-206EA 65.65 USD InStock
89059-206 89059-210 89059-208
Air Sampling Filter Screens, Ace Glass Incorporated
Manifolds
Filter screen made up of PTFE with 6.4 mm mesh is designed for use on the funnel end of air sampling canes.
The screen is held on to the bottom of the funnel by a stainless steel yoke retainer and three springs. Screen size is 152 mm.
Ordering information: Available complete with screen and retainer, or those component items may be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty