Would you like to log in again?
Midget Impinger, Ace-Threds, Sintered Glass Filter, Ace Glass Incorporated
Supplier: Ace Glass
7533-13 7533-18 7533-11 7533-28 7533-19 7533-27
89059-328EA 200.14 USD InStock
89059-328 89059-338 89059-326 89059-334 89059-332 89059-340
Midget Impinger, Ace-Threds, Sintered Glass Filter, Ace Glass Incorporated
Samplers Air Samplers
This piece is used for sampling air volumes at low jet velocity
Bottle joint is reinforced [ST] 24/40 outer. Tube uses #7 “Ace-Safe” removable hose connections for easy connect/disconnect. Nozzle has sintered glass filter, Porosity A (145-175 microns) or Porosity B (70-100 micron). Bottle has encircling graduations from 0 to 25mL or 0 to 30mL in 5mL divisions. Fabricated from borosilicate glass.
Ordering information: Complete item consists of bottle, tube, connectors and bushings. Items can also be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty