Would you like to log in again?
Midget Impinger, Ace Glass
Supplier: Ace Glass
7544-35
89059-386EA 228.22 USD InStock
89059-386
Midget Impinger, Ace Glass
Samplers Air Samplers
This unique impinger is designed to be more economical, when connecting in series, by eliminating the need for connecting tubes
The spherical 12/5 joints are perpendicular and come directly off the bottle 180° apart. The screw cap on the the bottle has a PTFE liner. Each bottle has one ball and one socket joint. The bottle has encircling graduations from 0 to 25 mL, or 0 to 30 mL in 5mL divisions. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty