Would you like to log in again?
Joints, Inner, Full Length, Ace Glass Incorporated
Supplier: Ace Glass
7565-27 7565-71 7565-52 7565-47 7565-57 7565-67 7565-72 7565-32 7565-62 7565-42 7565-07 7565-37 7565-17 7565-77 7565-12 7565-22
89059-448EA 18.32 USD InStock
89059-448 89059-458 89059-468 89059-446 89059-456 89059-466 89059-454 89059-476 89059-464 89059-474 89059-452 89059-462 89059-472 89059-450 89059-460 89059-470
Joints, Inner, Full Length, Ace Glass Incorporated
NS Joint Sleeves and Connectors Joint Sleeves
Standard taper inner ground joints are designed for use with matching outer members of the same size, such as 89059-456, 89059-492, 89059-520, 89060-016 and 89060-042
They can also be used with half joints, such as 89059-546 and 89059-578. All joints are manufactured from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty