Would you like to log in again?
Inner Joints, Full Length, Ace Glass Incorporated
Supplier: Ace Glass
7566-58 7566-18 7566-68 7566-33 7566-63 7566-21 7566-13 7566-48 7566-53 7566-26 7566-75 7566-28 7566-73 7566-38 7566-43 7566-23 7566-08
89084-180EA 600.06 USD InStock
89084-180 89059-508 89059-506 89059-504 89059-502 89059-500 89059-478 89059-488 89059-498 89059-486 89059-496 89059-484 89059-494 89059-482 89059-492 89059-480 89059-490
Inner Joints, Full Length, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The tubing of these full length inner joints is the same diameter as the reinforcing rings on the joint.
  • Reduced joint strain
  • Less tendency toward lubrication loss
  • Reinforcement rings instead of a heavy wall
Strain on the joint is reduced from expansion due to temperature increase. Properly clamped, the joint tends to remain cooler, with less tendency toward lubrication loss. Very heavy walls are not desirable for thermal shock resistance, but are desirable for resistance to mechanical pressure and shock. Therefore, providing reinforcement rings instead of a heavy wall results in a better, more balanced design. No holdup below the joint makes this an ideal joint for flask necks and receivers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty