Would you like to log in again?
Rodaviss® Loosening Ring, Polyamide, Ace Glass Incorporated
Supplier: Ace Glass
7618-07 7618-03 7618-05 7618-13 7618-09
89059-756EA 4.28 USD InStock
89059-756 89059-754 89059-752 89059-750 89059-760
Rodaviss® Loosening Ring, Polyamide, Ace Glass Incorporated
Support Rings
Loosening rings are designed to assist in freeing frozen joints.
  • Loosening rings
  • Made of polyamide
Simply insert between top of cap and ring on shank of inner member. Unscrew cap and inner member will release.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty