Would you like to log in again?
Cap, GL, with Hole, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7621-15 7621-25 7621-04 7621-08 7621-22
89059-778EA 6.79 USD InStock
89059-778 89059-776 89059-774 89059-782 89059-780
Cap, GL, with Hole, Ace Glass Incorporated
Closures Bottle Closures
This open top red polybutylene teraphthalate (PBT) cap is for use with GL threads
When used with a hose connection and appropriate sealing ring, this cap will accommodate tubing of the same approximate diameter. Temperature range is –45ºC to 180ºC.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty