Would you like to log in again?
Moisture Test Receiver, Phosphorous Determination, Ace Glass Incorporated
Supplier: Ace Glass
7729-10
89060-334EA 426.28 USD InStock
89060-334
Moisture Test Receiver, Phosphorous Determination, Ace Glass Incorporated
Distillation Components Distillation Receivers
Used in phosphorus determination
Receiver capacity is 5 mL, graduated in 0.05 mL subdivisions. Upper joint is a standard taper 24/40, side arm joint, stadnard taper 45/50. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty