Would you like to log in again?
Vacuum Filtration Tube, 3 Neck with PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7761-26 7761-17 7761-37 7761-27 7761-36 7761-16
89060-380EA 554.87 USD InStock
89060-380 89060-400 89060-402 89060-386 89060-382 89060-384
Vacuum Filtration Tube, 3 Neck with PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
In addition to collecting and filtering, this vessel may be used as a crystallizer.
  • 2 mm bore PTFE stopcock on 20 and 80 mL sizes
  • 3 mm bore PTFE on 250 mL size
  • Two [ST] outer joints
  • Two [ST] inner joints
  • With fritted disc
This 3-neck vacuum filtration tube comes with a 2 mm bore PTFE stopcock on the 20 and 80 mL sizes, and a 3 mm bore PTFE on the 250 mL size. In addition to collecting and filtering, this vessel may be used as a crystallizer. With two standard taper 14/20 or standard taper 24/25 outer joints and two standard taper 14/35 or standard taper 24/40 inner. With fritted disc (Porosity C = 25-50 micron; Porosity D = 10-20 micron).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty