Would you like to log in again?
Glass Storage Tube Cap with Glass Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7797-07 7797-05 7797-16
89060-444EA 146.19 USD InStock
89060-444 89060-440 89060-442
Glass Storage Tube Cap with Glass Stopcock, Ace Glass Incorporated
Closures Test/Sample Tube Closures
These caps are for use with storage tubes
With 2 mm bore stopcock for evacuating or introduction of inert gases. The standard taper 14/35 or standard taper 24/40 joints are used when it is necessary to evacuate completely. The standard taper 14/20 joint is used when there is more concern about grease entering the system. Tube outlet is 8 mm O.D. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty