Would you like to log in again?
Adapter, 115 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7803-25 7803-12
89060-452EA 84.12 USD InStock
89060-452 89060-450
Adapter, 115 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This 115 degree angle adapter is fabricated from borosilicate glass.
  • 115° angle bend
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Fabricated from borosilicate glass
Adapter has an outer standard taper joint at the top and an inner standard taper joint at the bottom.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty