Would you like to log in again?
Adapter, Straight, Ace Glass Incorporated
Supplier: Ace Glass
7805-12
89060-454EA 114.67 USD InStock
89060-454
Adapter, Straight, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features two standard taper outer joints, one with a drip tube.
  • Top and bottom [ST] outer joints
  • Drip tube
  • Fabricated from borosilicate glass
Adapter has two standard taper 14/20 outer joints, one with a 7mm drip tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty