Would you like to log in again?
Photochemical Quartz Double-Walled Immersion Well, Ace Glass Incorporated
Supplier: Ace Glass
7854-27 7854-28 7854-26 7854-25
89060-588EA 2231.77 USD InStock
89060-588 89060-594 89060-590 89060-592
Photochemical Quartz Double-Walled Immersion Well, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Double-walled, quartz immersion well for use with certain U.V
  • Well has a standard taper 60/40 inner joint
  • Length above joint is 76 mm
Ace lamps. With inlet and outlet tubes for cooling. A small diameter inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty