Would you like to log in again?
Photochemical Borosilicate Glass Double-Walled Immersion Well, Ace Glass Incorporated
Supplier: Ace Glass
7857-11 7857-10 7857-06 7857-05
89061-226EA 1132.69 USD InStock
89061-226 89061-228 89061-232 89061-230
Photochemical Borosilicate Glass Double-Walled Immersion Well, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Double-walled, borosilicate glass immersion well for use with U.V
  • Well has a standard taper 60/40 inner joint
  • Outer jacket is 53 mm O.D.
  • Length above joint is 76 mm
Optional PTFE tube attached to a glass tube and secured wioth a #7 Ace-Thred bushing. PTFE tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet.
lamps. With inlet and outlet tubes for cooling. A small diameter inlet tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty