Would you like to log in again?
Chemraz® O-rings, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7859-520 7859-540 7859-502 7859-517 7859-512 7859-542 7859-571 7859-530 7859-510 7859-576 7859-522 7859-574 7859-513 7859-579 7859-503 7859-526 7859-528 7859-539 7859-578 7859-519 7859-544 7859-518 7859-572 7859-575 7859-545 7859-538 7859-506 7859-516 7859-504 7859-536 7859-501 7859-505 7859-547 7859-527 7859-508 7859-529 7859-524 7859-548 7859-550 7859-570 7859-514 7859-573 7859-515 7859-532 7859-534 7859-521 7859-507 7859-511 7859-546
89061-324EA 310.79 USD InStock
89061-324 89061-322 89061-328 89061-326 89061-280 89061-284 89061-282 89061-288 89061-320 89061-286 89061-314 89061-312 89061-278 89061-318 89061-316 89061-350 89061-272 89061-270 89061-310 89061-354 89061-276 89061-352 89061-274 89061-346 89061-302 89061-268 89061-300 89061-344 89061-306 89061-348 89061-304 89061-308 89061-262 89061-260 89061-266 89061-342 89061-264 89061-340 89061-336 89061-258 89061-334 89061-338 89061-290 89061-294 89061-292 89061-332 89061-298 89061-330 89061-296
Chemraz® O-rings, Ace Glass Incorporated
Closures Bottle Closures
Known as "the white o-ring", Chemrez is a white, perfluoroelastomer compound with high temperature and chemical resistance
Compared with other elastomers, Chemraz is normally more resistant to swelling and embrittlement, and will retain these properties for a longer period of time. Compared with metal seals, Chemraz is easily installed and conforms to the sealing surface despite irregularities that may occur due to improper assembly or wear. Compared with PTFE seals, Chemraz is not likely to creep or cold flow.
CHEMRAZ o-rings retain their elastic properties longer in harsh chemical environments at temperatures from -30°C to 220°C. Recommended for use in a variety of hot, corrosive environments.
It has all the properties and inertness of PTFE. Great for Pharmaceutical or food applications where high purity is a must.
Ordering information: Chemraz is a registered trademark of Green, Tweed and Co.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty