Would you like to log in again?
Photochemical Unjacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Supplier: Ace Glass
7863-20 7863-36 7863-38 7863-40 7863-16 7863-18
89061-366EA 879.33 USD InStock
89061-366 89060-576 89084-400 89060-574 89060-580 89061-364
Photochemical Unjacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Reaction vessel fabricated of borosilicate glass to accommodate the immersion wells only, With #50 Ace-Thred at top which accepts a 7506-14 bushing and FETFE O-Ring to form a compression seal with the immersion well, one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one #7-Ace-Thred side arm for thermometer
Bottom of reactor is flat to allow use of magnetic stirrer. Volumes indicated are total volumes. Working volume in reactive area of lamp is approximately 40-50% of total volume. Complete unit consists of vessel, sparger tube, PTFE stir bar, threaded nylon thermometer bushing, #50 nylon bushing and three meters of 4.8 mm (3/16") I.D. PTFE tubing.
Ordering information: Complete Units Include:
Vessel
89060-570 Sparger Tube
89077-842 PTFE Stir Bar, 38mm X 8mm
89051-122 #7 Nylon Thermometer Bushing
89059-230 #50 Nylon Bushing
PTFE Tubing, 4.8 mm I.D. x 3 m
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty