Would you like to log in again?
Photochemical Reaction Vessel, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7841-15 7841-09 7841-19 7841-10 7841-03 7841-06 7841-16 7841-04 7841-14 7841-05
89061-368EA 1079.64 USD InStock
89061-368 89060-566 89060-568 89060-578 89061-372 89060-572 89061-370 89060-564 89060-570 89061-362
Photochemical Reaction Vessel, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Reaction vessel constructed of borosilicate glass to accommodate the immersion wells
With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one- #7 Ace-Thred side arm for thermometer. Center joint is standard taper 60/40. Bottom of reactor is flat to allow use of magnetic stirrer. Volumes indicated are total volumes. Volume in reactive area of lamp is approx. 40-50% of total volume.
Ordering information: Complete unit consists of vessel, sparger tube, PTFE stir bar, threaded nylon bushing and three meters of 4.8 mm (3/16") I.D. PTFE tubing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty