Would you like to log in again?
Microscale Photochemical Reaction Assembly, Ace Glass Incorporated
Supplier: Ace Glass
7880-62 7880-20 7880-15 7880-60 7880-12
89061-390EA 131.77 USD InStock
89061-390 89061-394 89061-392 89061-398 89061-396
Microscale Photochemical Reaction Assembly, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Reactor body is fabricated of borosilicate glass with #25 Ace-Thred at top, standard taper 14/10 outer side joint externally threaded to allow an O-Ring cap seal connection with mating inner joint, and porosity E (4-8micron) filter disc at bottom for use as a gas inlet to remove oxygen from the solution and/or for agitation (mixing)
Bottom hose connection is size E. Immersion well is either quartz or borosilicate, 11 mm I.D., with water jacket. Well can also be used to flow filter solutions in front of lamp to attain desired wavelengths. Well is assembled by insertion into #25 Ace-Thred and securing with an O‑Ring compression seal. This type connection allows volume adjusting by raising or lowering well inside reactor, volume range 5-15 mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty