Would you like to log in again?
Cannon-Fenske Viscometer, Ace Glass Incorporated
Supplier: Ace Glass
7988-21 7988-25 7988-43 7988-13 7988-17 7988-51 7988-07 7988-09 7988-11 7988-19 7988-45 7988-37 7988-15 7988-29 7988-53 7988-41 7988-55 7988-23 7988-35 7988-33 7988-47 7988-39 7988-49 7988-59 7988-03 7988-57 7988-05 7988-27
89061-544EA 249.47 USD InStock
89061-544 89061-566 89061-588 89061-542 89061-564 89061-586 89061-548 89061-546 89061-568 89061-580 89061-540 89061-584 89061-562 89061-560 89061-582 89061-556 89061-578 89061-534 89061-554 89061-576 89061-538 89061-536 89061-558 89061-570 89061-552 89061-574 89061-572 89061-550
Cannon-Fenske Viscometer, Ace Glass Incorporated
Viscometers Capillary Viscometers
For use in obtaining kinematic viscosities
Directions for use are published in ASTM Standards on Petroleum Products and Lubricants, D 445. Viscosities can be measured with a precision of ±0.2% quickly and easily. A sample of approximately 7 mL is necessary. Offered calibrated with certificate or uncalibrated.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty