Would you like to log in again?
Sublimation Adapter, Ace Glass Incorporated
Supplier: Ace Glass
8015-20 8015-15
89061-622EA 352.2 USD InStock
89061-622 89061-624
Sublimation Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Unique device for carrying out sublimation directly in a round bottom reaction flask.
  • Coolant connection at top
  • Side vacuum connection
  • 1:5 PTFE 2 mm bore stopcock
  • [ST] joint for use with 250 mL or 500 mL flasks
Simply remove condenser, stirrer, etc. from flask joint, insert sublimation adapter and begin. Coolant connection at top and side vacuum connection are 9.5 mm (3/8") O.D. Vacuum connection is controlled by a 1:5 PTFE 2 mm bore stopcock. Adapter is supplied with a standard taper 24/40 joint for use with 250 mL or 500 mL flasks. Other joint sizes or capacities are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty