Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vacuum Sublimation Apparatus, Large Scale, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8027-25 8027-14 8027-09 8027-02 8027-05 8027-07
89061-654EA 847.45 USD InStock
89061-654 89061-664 89061-658 89061-656 89061-662 89061-660
Vacuum Sublimation Apparatus, Large Scale, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This large-scale 2L sublimation apparatus is constructed of heavy wall glass.

  • Unique design offers both a sublimation apparatus as well as a two-liter reaction kettle
  • Unit will sublime approximately 100 gms of solid sample
  • Constructed of heavy-wall glass

Unit will sublime approximately 100 gms of solid sample. Unique design offers both a sublimation apparatus as well as a two-liter reaction kettle. Cold finger tube has a standard taper 34/35 joint which positions in the center neck and is held by means of a plastic bushing to a 300 mL condensate flask. The wide flange opening of the kettle allows for easy removal and access to the condenser flask. Side joints are standard taper 24/40. Kettle flange has an o-ring grove for a CAPFE O-Ring, (supplied) for easy grease free seal. Head and kettle are joined with a quick release clamp.

Ordering information: Complete system has all the components listed below. 89061-662 stopcock adapter uses size "C" (⅜") hose connections. 89061-658 condenser tube uses size A (5/16") hose connections.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty