Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Complete Precision Trubore® Stirrer Assembly, Ace Glass Incorporated
Supplier: Ace Glass
8041-38 8041-40 8041-11 8041-15 8041-19 8041-30 8041-20 8041-34
89061-754EA 374.97 USD InStock
89061-754 89061-742 89061-752 89061-740 89061-746 89061-744 89061-748 89061-750
Complete Precision Trubore® Stirrer Assembly, Ace Glass Incorporated
Stirrers Overhead Stirrers
Precision Trubore® Stirrer Assembly consisting of 10 mm I.D. bearing, bushing, precision ground, button type glass shaft and blade.
Available with either glass or PTFE stir blade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty